1-Bromo-3,5-bis(methylthio)benzene 95%

Supplier: AOBChem USA

Warning

25579-1G 25579-25G 25579-5G 25579-500MG
76885-738EA 114.58 USD
76885-738 76885-740 76885-742 77645-480
1-Bromo-3,5-bis(methylthio)benzene 95%
1-Bromo-3,5-bis(methylthio)benzene
Formula: C₈H₉BrS₂
MW: 249.191 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD33403707
CAS Number: 149428-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR