2-(2-Methylphenyl)benzonitrile 95%

Supplier: AOBChem USA

Warning

25515-1G 25515-25G 25515-5G 25515-500MG
76885-714EA 117.76 USD
76885-714 76885-716 76885-718 77649-544
2-(2-Methylphenyl)benzonitrile 95%
2-(2-Methylphenyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₁N
Storage Temperature: Refrigerator
MDL Number: MFCD05980950
CAS Number: 157366-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR