1-(2-Chloropyridin-3-yl)ethanol ≥97%

Supplier: AOBChem USA

Warning

25355-1G 25355-25G 25355-5G
76885-642EA 66.84 USD
76885-642 76885-644 76885-646
1-(2-Chloropyridin-3-yl)ethanol ≥97%
1-(2-Chloropyridin-3-yl)ethanol
Formula: C₇H₈ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD16037360
CAS Number: 131674-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR