1-Bromo-4-methoxy-2-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

25244-1G 25244-25G 25244-500MG 25244-5G
76885-622EA 171.87 USD
76885-622 76885-624 76885-626 76885-628
1-Bromo-4-methoxy-2-(methoxymethyl)benzene ≥97%
1-Bromo-4-methoxy-2-(methoxymethyl)benzene
Formula: C₉H₁₁BrO₂
MW: 231.086 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21193511
CAS Number: 94527-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR