1,3-Dibromo-5-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

25101-1G 25101-25G 25101-5G
76885-584EA 62.6 USD
76885-584 76885-586 76885-588
1,3-Dibromo-5-(methoxymethoxy)benzene ≥97%
1,3-Dibromo-5-(methoxymethoxy)benzene
Formula: C₈H₈Br₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD27928576
CAS Number: 770718-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR