1-(Methoxymethoxy)-3,5-bis(trifluoromethyl)-benzene ≥97%

Supplier: AOBChem USA

Warning

24824-25G 24824-5G
76885-558EA 466.8 USD
76885-558 76885-560
1-(Methoxymethoxy)-3,5-bis(trifluoromethyl)-benzene ≥97%
1-(Methoxymethoxy)-3,5-bis(trifluoromethyl)-benzene
Formula: C₁₀H₈F₆O₂
MW: 274.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31619416
CAS Number: 1032638-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR