(5,6-Dimethoxypyridin-3-yl)boronic acid ≥98%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Dimethoxypyridine-5-boronic acid

24746-1G 24746-25G 24746-500MG 24746-5G
76885-546EA 290.69 USD
76885-546 76885-548 76885-550 76885-552
(5,6-Dimethoxypyridin-3-yl)boronic acid ≥98%
(5,6-Dimethoxypyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₄
MW: 182.97 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18254414
CAS Number: 1346526-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR