1-Ethyl-3-(methylthio)benzene ≥97%

Supplier: AOBChem USA

Warning

24721-1G 24721-25G 24721-500MG 24721-5G
76885-530EA 260.99 USD
76885-530 76885-532 76885-534 76885-536
1-Ethyl-3-(methylthio)benzene ≥97%
1-Ethyl-3-(methylthio)benzene
Formula: C₉H₁₂S
MW: 152.257 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11617784
CAS Number: 66794-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR