1-Bromo-3,5-Difluoro-4-(Methylsulfanyl)Benzene ≥97%

Supplier: AOBChem USA

Warning

24640-25G 24640-5G
76885-514EA 310.85 USD
76885-514 76885-516
1-Bromo-3,5-Difluoro-4-(Methylsulfanyl)Benzene ≥97%
1-Bromo-3,5-Difluoro-4-(Methylsulfanyl)Benzene
Formula: C₇H₅BrF₂S
Storage Temperature: Ambient
MDL Number: MFCD22628201
CAS Number: 648905-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR