1-(4-Bromo-2-chlorobenzyl)pyrrolidine hydrochloride ≥97%

Supplier: AOBChem USA

Warning

24510-1G 24510-25G 24510-500MG 24510-5G
76885-482EA 147.47 USD
76885-482 76885-484 76885-486 76885-488
1-(4-Bromo-2-chlorobenzyl)pyrrolidine hydrochloride ≥97%
1-(4-Bromo-2-chlorobenzyl)pyrrolidine hydrochloride
Formula: C₁₁H₁₄BrCl₂N
MW: 311.046 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22543715
CAS Number: 1394291-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR