4-Bromo-2-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Bromo-2-methylphenyl)methanol

24446-25G 24446-5G
76885-472EA 212.18 USD
76885-472 76885-474
4-Bromo-2-methylbenzyl alcohol ≥97%
4-Bromo-2-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO
Storage Temperature: Refrigerator
MDL Number: MFCD11847398
CAS Number: 17100-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR