2,4-Bis(trifluoromethyl)-6-bromophenol ≥95% research grade

Supplier: AOBChem USA

Warning

24408-1G 24408-25G 24408-500MG 24408-5G
76885-454EA 350.1 USD
76885-454 76885-456 76885-458 76885-460
2,4-Bis(trifluoromethyl)-6-bromophenol ≥95% research grade
2,4-Bis(trifluoromethyl)-6-bromophenol
Formula: C₈H₃BrF₆O
Storage Temperature: Refrigerator
MDL Number: MFCD22628192
CAS Number: 1414870-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR