1,5-Dibromo-2,3,4-trifluorobenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
24339-25G
24339-5G
76885-420EA
364.95
USD
76885-420
76885-422
1,5-Dibromo-2,3,4-trifluorobenzene ≥97%
1,5-Dibromo-2,3,4-trifluorobenzene
Formula:
C₆HBr₂F₃ MW: 289.875 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28505052 CAS Number: 17299-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...