2-(4-Bromophenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(4-Bromophenyl)-2-propanol

24190-25G 24190-5G
76885-396EA 481.65 USD
76885-396 76885-398
2-(4-Bromophenyl)propan-2-ol ≥97%
2-(4-Bromophenyl)propan-2-ol
Formula: C₉H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD11870096
CAS Number: 2077-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR