(3-Chloropyridin-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

24119-1G 24119-25G 24119-5G
76885-386EA 93.36 USD
76885-386 76885-388 76885-390
(3-Chloropyridin-4-yl)methanol ≥97%
(3-Chloropyridin-4-yl)methanol
Formula: C₆H₆ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD09910243
CAS Number: 79698-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR