2-(4-Fluorophenyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

23922-25G 23922-5G 23922-1G
76885-364EA 204.76 USD
76885-364 76885-366 77641-410
2-(4-Fluorophenyl)benzaldehyde ≥97%
2-(4-Fluorophenyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₉FO
Storage Temperature: Ambient
MDL Number: MFCD01631854
CAS Number: 192863-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR