1,4-Dibromo-2-isopropoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

23759-25G 23759-5G
76885-314EA 488.02 USD
76885-314 76885-316
1,4-Dibromo-2-isopropoxybenzene ≥97% research grade
1,4-Dibromo-2-isopropoxybenzene
Formula: C₉H₁₀Br₂O
Storage Temperature: Ambient
MDL Number: MFCD22543671
CAS Number: 1369842-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR