Di-tert-butyl piperazine-1,4-dicarboxylate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1,4-Bis(tert-butoxycarbonyl)piperazine

23646-25G 23646-5G
76885-280EA 204.76 USD
76885-280 76885-282
Di-tert-butyl piperazine-1,4-dicarboxylate ≥97% research grade
Di-tert-butyl piperazine-1,4-dicarboxylate
Formula: C₁₄H₂₆N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD01435774
CAS Number: 76535-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR