1,3-Dibromo-2-(methylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

23630-25G 23630-5G
76885-270EA 524.09 USD
76885-270 76885-272
1,3-Dibromo-2-(methylthio)benzene ≥95%
1,3-Dibromo-2-(methylthio)benzene
Formula: C₇H₆Br₂S
Storage Temperature: Refrigerator
MDL Number: MFCD22543664
CAS Number: 301151-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR