1-Fluoro-4-Methyl-2-(Methylsulfanyl)Benzene ≥97%

Supplier: AOBChem USA

Warning

23600-1G 23600-25G 23600-5G
76885-258EA 64.72 USD
76885-258 76885-260 76885-262
1-Fluoro-4-Methyl-2-(Methylsulfanyl)Benzene ≥97%
1-Fluoro-4-Methyl-2-(Methylsulfanyl)Benzene
Formula: C₈H₉FS
MW: 156.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527259
CAS Number: 1370025-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR