6-[4-(Trifluoromethoxy)phenyl]-3-pyridinecarbaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-(4-(Trifluoromethoxy)phenyl)pyridine-5-carboxaldehyde

23565-1G 23565-25G 23565-500MG 23565-5G
76885-244EA 231.28 USD
76885-244 76885-246 76885-248 76885-250
6-[4-(Trifluoromethoxy)phenyl]-3-pyridinecarbaldehyde ≥95%
6-[4-(Trifluoromethoxy)phenyl]-3-pyridinecarbaldehyde
Formula: C₁₃H₈F₃NO₂
MW: 267.203 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04115436
CAS Number: 851069-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR