1-(4-Bromobenzyl)pyrrolidine ≥97%

Supplier: AOBChem USA

Warning

23461-25G 23461-5G
76885-232EA 418 USD
76885-232 76885-234
1-(4-Bromobenzyl)pyrrolidine ≥97%
1-(4-Bromobenzyl)pyrrolidine
Formula: C₁₁H₁₄BrN
MW: 240.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06080622
CAS Number: 4897-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR