2,3-Dichloro-6-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

23381-1G 23381-25G 23381-5G
76885-206EA 79.57 USD
76885-206 76885-208 76885-210
2,3-Dichloro-6-fluorophenol ≥97%
2,3-Dichloro-6-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₃Cl₂FO
Storage Temperature: Ambient
CAS Number: 886497-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR