1-Bromo-4-methoxy-2-methyl-3-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

23264-1G 23264-25G 23264-500MG 23264-5G
76885-182EA 203.7 USD
76885-182 76885-184 76885-186 76885-188
1-Bromo-4-methoxy-2-methyl-3-nitrobenzene ≥97%
1-Bromo-4-methoxy-2-methyl-3-nitrobenzene
Formula: C₈H₈BrNO₃
MW: 246.058 g/mol
Storage Temperature: Ambient
CAS Number: 85598-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR