2-Isopropoxy-1-naphthaldehyde ≥97%

Supplier: AOBChem USA

Warning

23178-1G 23178-25G 23178-5G
76885-156EA 74.27 USD
76885-156 76885-158 76885-160
2-Isopropoxy-1-naphthaldehyde ≥97%
2-Isopropoxy-1-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD02629634
CAS Number: 885-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR