1,5-Dichloro-2,4-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

23024-1G 23024-25G 23024-5G
76885-110EA 84.88 USD
76885-110 76885-112 76885-114
1,5-Dichloro-2,4-difluorobenzene ≥95%
1,5-Dichloro-2,4-difluorobenzene
Formula: C₆H₂Cl₂F₂
Storage Temperature: Refrigerator
MDL Number: MFCD11226309
CAS Number: 2253-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR