1-Fluoro-4,5-dimethoxy-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

23001-1G 23001-500MG 23001-5G
76885-098EA 528.33 USD
76885-098 76885-100 76885-102
1-Fluoro-4,5-dimethoxy-2-iodobenzene ≥97%
1-Fluoro-4,5-dimethoxy-2-iodobenzene
Formula: C₈H₈FIO₂
Storage Temperature: Refrigerator
MDL Number: MFCD19440150
CAS Number: 900175-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR