1,3-Dibromo-2-(trimethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

22955-1G 22955-25G 22955-500MG 22955-5G
76885-086EA 182.48 USD
76885-086 76885-088 76885-090 76885-092
1,3-Dibromo-2-(trimethylsilyl)benzene ≥95%
1,3-Dibromo-2-(trimethylsilyl)benzene
Formula: C₉H₁₂Br₂Si
Storage Temperature: Ambient
MDL Number: MFCD32669955
CAS Number: 363598-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR