(4'-formyl-[1,1'-biphenyl]-4-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

22903-1G 22903-25G 22903-500MG 22903-5G
76885-072EA 296 USD
76885-072 76885-074 76885-076 76885-078
(4'-formyl-[1,1'-biphenyl]-4-yl)boronic acid ≥95%
(4'-formyl-[1,1'-biphenyl]-4-yl)boronic acid
Formula: C₁₃H₁₁BO₃
MW: 226.036 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08701746
CAS Number: 868046-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR