1,7-Dimethyl-1H-indazole-5-boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Borono-1,7-dimethyl-1H-indazole

22440-1G 22440-25G 22440-500MG 22440-5G
76885-020EA 263.11 USD
76885-020 76885-022 76885-024 76885-026
1,7-Dimethyl-1H-indazole-5-boronic acid ≥97%
1,7-Dimethyl-1H-indazole-5-boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BN₂O₂
MW: 190.007 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11226596
CAS Number: 1310383-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR