2,3-Dichloro-6-bromobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

22368-1G 22368-25G 22368-500MG 22368-5G
76885-004EA 320.4 USD
76885-004 76885-006 76885-008 76885-010
2,3-Dichloro-6-bromobenzoic acid ≥97%
2,3-Dichloro-6-bromobenzoic acid
Formula: C₇H₃BrCl₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD07368091
CAS Number: 887584-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR