2,4-Dibromo-1-(methoxymethoxy)benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
22323-1G
22323-25G
22323-500MG
22323-5G
76884-992EA
194.15
USD
76884-992
76884-994
76884-996
76884-998
2,4-Dibromo-1-(methoxymethoxy)benzene ≥97%
2,4-Dibromo-1-(methoxymethoxy)benzene
Formula:
C₈H₈Br₂O₂ MW: 295.956 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD26399858 CAS Number: 21571-52-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...