2,3-Difluoro-4-iodophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

22285-1G 22285-500MG 22285-5G
76884-984EA 560.16 USD
76884-984 76884-986 76884-988
2,3-Difluoro-4-iodophenylboronic acid ≥97%
2,3-Difluoro-4-iodophenylboronic acid
Formula: C₆H₄BF₂IO₂
Storage Temperature: Ambient
MDL Number: MFCD07644468
CAS Number: 1257793-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR