2,3-Dichloro-4-hydroxypyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Dichloropyridin-4-ol

22087-1G 22087-25G 22087-5G
76884-920EA 108.22 USD
76884-920 76884-922 76884-924
2,3-Dichloro-4-hydroxypyridine ≥97%
2,3-Dichloro-4-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₃Cl₂NO
MW: 163.989 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16610774
CAS Number: 1174047-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR