1-Bromo-3-propoxy-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

21958-1G 21958-25G 21958-500MG 21958-5G
76884-904EA 173.99 USD
76884-904 76884-906 76884-908 76884-910
1-Bromo-3-propoxy-5-(trifluoromethyl)benzene ≥97%
1-Bromo-3-propoxy-5-(trifluoromethyl)benzene
Formula: C₁₀H₁₀BrF₃O
MW: 283.085 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29043916
CAS Number: 1880282-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR