1-(Benzyloxy)-3-bromo-5-trifluoromethylbenzene ≥97%

Supplier: AOBChem USA

Warning

21953-1G 21953-25G 21953-500MG 21953-5G
76884-888EA 159.14 USD
76884-888 76884-890 76884-892 76884-894
1-(Benzyloxy)-3-bromo-5-trifluoromethylbenzene ≥97%
1-(Benzyloxy)-3-bromo-5-trifluoromethylbenzene
Formula: C₁₄H₁₀BrF₃O
MW: 331.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09955461
CAS Number: 130723-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR