1,4-Dibromo-2,5-dichlorobenzene ≥95% research grade

Supplier: AOBChem USA

Warning

21894-25G 21894-5G
76884-874EA 297.06 USD
76884-874 76884-876
1,4-Dibromo-2,5-dichlorobenzene ≥95% research grade
1,4-Dibromo-2,5-dichlorobenzene
Formula: C₆H₂Br₂Cl₂
MW: 304.79 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00182443
CAS Number: 4571-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR