2-(3,5-Dimethoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

21857-25G 21857-5G
76884-866EA 424.36 USD
76884-866 76884-868
2-(3,5-Dimethoxyphenyl)-1,3-dioxolane ≥97%
2-(3,5-Dimethoxyphenyl)-1,3-dioxolane
Formula: C₁₁H₁₄O₄
MW: 210.226 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06208952
CAS Number: 140464-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR