(4-Bromo-3,5-dimethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

21738-1G 21738-25G 21738-5G
76884-840EA 66.84 USD
76884-840 76884-842 76884-844
(4-Bromo-3,5-dimethylphenyl)methanol ≥97%
(4-Bromo-3,5-dimethylphenyl)methanol
Formula: C₉H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD00277869
CAS Number: 27006-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR