1H-Indazole-5-carbonitrile ≥97%

Supplier: AOBChem USA

Warning

21693-25G 21693-5G
76884-832EA 587.74 USD
76884-832 76884-834
1H-Indazole-5-carbonitrile ≥97%
1H-Indazole-5-carbonitrile
Formula: C₈H₅N₃
MW: 143.1454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973397
CAS Number: 74626-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR