2,6-Dibromo-4-nitrotoluene ≥95%

Supplier: AOBChem USA

Warning

21265-25G 21265-5G
76884-768EA 201.58 USD
76884-768 76884-770
2,6-Dibromo-4-nitrotoluene ≥95%
2,6-Dibromo-4-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD00151807
CAS Number: 110127-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR