2,4-Difluorophenetole ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1-Ethoxy-2,4-difluorobenzene

21239-1G 21239-25G 21239-5G
76884-758EA 101.85 USD
76884-758 76884-760 76884-762
2,4-Difluorophenetole ≥97% research grade
2,4-Difluorophenetole
Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11846411
CAS Number: 348-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR