1-Fluoro-4-(1-methylethoxy)-benzene ≥97%

Supplier: AOBChem USA

Warning

21142-1G 21142-25G 21142-5G 21142-500MG
76884-730EA 76.39 USD
76884-730 76884-732 76884-734 77652-474
1-Fluoro-4-(1-methylethoxy)-benzene ≥97%
1-Fluoro-4-(1-methylethoxy)-benzene
Formula: C₉H₁₁FO
MW: 154.181 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18447616
CAS Number: 459-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR