(4-Fluoro-1H-indazol-6-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

21113-1G 21113-25G 21113-500MG 21113-5G
76884-714EA 409.51 USD
76884-714 76884-716 76884-718 76884-720
(4-Fluoro-1H-indazol-6-yl)boronic acid ≥97%
(4-Fluoro-1H-indazol-6-yl)boronic acid
Formula: C₇H₆BFN₂O₂
MW: 179.944 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18383477
CAS Number: 1253911-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR