1-(1-Ethoxyethyl)-4-iodo-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

21079-25G 21079-5G
76884-694EA 360.71 USD
76884-694 76884-696
1-(1-Ethoxyethyl)-4-iodo-1H-pyrazole ≥97%
1-(1-Ethoxyethyl)-4-iodo-1H-pyrazole

Precaución: for Research Use Only

Formula: C₇H₁₁IN₂O
Storage Temperature: Ambient
MDL Number: MFCD22123601
CAS Number: 575452-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR