2-Acetamido-5-aminopyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-(Acetylamino)-5-aminopyridine
20843-25G
20843-5G
76884-664EA
240.83
USD
76884-664
76884-666
2-Acetamido-5-aminopyridine ≥97%
2-Acetamido-5-aminopyridine
Precaución: for Research Use Only
Formula:
C₇H₉N₃O MW: 151.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00160311 CAS Number: 29958-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...