1-Bromo-2-ethoxy-4-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

20758-25G 20758-5G
76884-646EA 233.4 USD
76884-646 76884-648
1-Bromo-2-ethoxy-4-fluorobenzene ≥95%
1-Bromo-2-ethoxy-4-fluorobenzene
Formula: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD03095002
CAS Number: 871717-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR