1-Bromo-2,4-difluoro-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

20684-1G 20684-25G 20684-500MG 20684-5G
76884-616EA 198.39 USD
76884-616 76884-618 76884-620 76884-622
1-Bromo-2,4-difluoro-3-methoxybenzene ≥97%
1-Bromo-2,4-difluoro-3-methoxybenzene
Formula: C₇H₅BrF₂O
MW: 223.015 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12924862
CAS Number: 221221-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR