1-Bromo-2,4-diethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

20569-1G 20569-25G 20569-500MG 20569-5G
76884-602EA 381.93 USD
76884-602 76884-604 76884-606 76884-608
1-Bromo-2,4-diethoxybenzene ≥97%
1-Bromo-2,4-diethoxybenzene
Formula: C₁₀H₁₃BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD18917793
CAS Number: 121550-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR