1-Bromo-4-chloro-3-(difluoromethyl)-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

20476-1G 20476-500MG 20476-5G
76884-594EA 528.33 USD
76884-594 76884-596 76884-598
1-Bromo-4-chloro-3-(difluoromethyl)-2-fluorobenzene ≥97%
1-Bromo-4-chloro-3-(difluoromethyl)-2-fluorobenzene
Formula: C₇H₃BrClF₃
MW: 259.451 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28733888
CAS Number: 943830-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR